missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ENG, Clone: 1E5., Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00002022-M26A.200uL
This item is not returnable.
View return policy
Specifications
| ENG | |
| Monoclonal | |
| Unconjugated | |
| In ascites fluid | |
| CD105/END/FLJ41744/HHT1/ORW/ORW1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Ascites |
| ELISA, Western Blot | |
| 1E5 | |
| Mouse monoclonal antibody raised against a full-length recombinant ENG. | |
| BC014271 | |
| ENG | |
| ENG (AAH14271, 27 a.a. ∼ 658 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2022 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction