Learn More
EPHB3 (M04), Mouse anti-Human, Clone: 2G8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPHB3.
Brand: Abnova H00002049-M04.100ug
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq
Sequence: AASLKVIASAQSGMSQPLLDRTVPDYTTFTTVGDWLDAIKMGRYKESFVSAGFASFDLVAQMTAEDLLRIGVTLAGHQKKILSSIQDMRLQMNQTLPVQSpecifications
| EPHB3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004443 | |
| EPHB3 | |
| EPHB3 (NP_004434, 899 a.a. ∼ 997 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 2G8 | |
| Mouse monoclonal antibody raised against a partial recombinant EPHB3. | |
| EPHB3 | |
| ETK2/HEK2/TYRO6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.