missing translation for 'onlineSavingsMsg'
Learn More
Learn More
establishment of cohesion 1 homolog 1 (S. cerevisiae), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ESCO1 protein.
Brand: Abnova H00114799-B01P.50ug
This item is not returnable.
View return policy
Description
ESCO1 belongs to a conserved family of acetyltransferases involved in sister chromatid cohesion (Hou and Zou, 2005 [PubMed 15958495]).[supplied by OMIM
Sequence: MVLPEDPKYALKKVDEIREMVDNDLGFQQAPLMCYSRTKTLLFISNDKKVVGCLIAEHIQWGYRVIEEKLPVIRSEEEKVRFERQKAWCCSTLPEPAICGISRIWVFSMMRRKKIASRMIECLRSNFIYGSYLSKEEIAFSDPTPDGKLFATQYCGTGQFLVYNFINGQNSTSpecifications
| establishment of cohesion 1 homolog 1 (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ESCO1 protein. | |
| ESCO1 | |
| A930014I12Rik/CTF/ECO1/EFO1/ESO1/KIAA1911/MGC105022 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC036943.1 | |
| ESCO1 | |
| ESCO1 (AAH36943.1, 1 a.a. ∼ 172 a.a) full-length human protein. | |
| 50 μg | |
| Endocrine/Metabolism | |
| 114799 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction