missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethylmalonic encephalopathy 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ETHE1 protein.
Brand: Abnova H00023474-B01P.50ug
This item is not returnable.
View return policy
Description
This gene encodes a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in this gene result in ethylmalonic encephalopathy
Sequence: MAEAVLRVARRQLSQRGGSGAPILLRQMFEPVSCTFTYLLGDRESREAVLIDPVLETAPRDAQLIKELGLRLLYAVNTHCHADHITGSGLLRSLLPGCQSVISRLSGAQADLHIEDGDSIRFGRFALETRASPGHTPGCVTFVLNDHSMAFTGDALLIRGCGRTDFQQGCAKTLYHSVHEKIFTLPGDCLIYPAHDYHGFTVSTVEEERTLNPRLTLSCEEFVKIMGNLNLPKPQQIDFAVPANMRCGVQTPTASpecifications
| ethylmalonic encephalopathy 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ETHE1 protein. | |
| ETHE1 | |
| HSCO/YF13H12 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 23474 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014297.3 | |
| ETHE1 | |
| ETHE1 (NP_055112.2, 1 a.a. ∼ 254 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction