Learn More
ETV6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant ETV6.
Brand: Abnova H00002120-A01.50uL
Description
This gene encodes an ETS family transcription factor. The product of this gene contains two functional domains: a N-terminal pointed (PNT) domain that is involved in protein-protein interactions with itself and other proteins, and a C-terminal DNA-binding domain. Gene knockout studies in mice suggest that it is required for hematopoiesis and maintenance of the developing vascular network. This gene is known to be involved in a large number of chromosomal rearrangements associated with leukemia and congenital fibrosarcoma. [provided by RefSeq
Sequence: MSETPAQCSIKQERISYTPPESPVPSYASSTPLHVPVPRALRMEEDSIRLPAHLRLQPIYWSRDDVAQWLKWAENEFSLRPIDSNTFEMNGKALLLLTKEDFRYRSPHSGDVLYELLQHILKQRKPRILFSPFFHPGNSIHTQPEVILHQNHEEDNCVQRTPRPSVDNVHHNPPTIELLHRSRSPITTNHRPSPDPEQRPLRSPLDNMIRRLSPAERAQGPRPHQENNHQESYPLSVSPMENNHCPASSESHPKPSSPRQESTRVIQLMPSPIMHPLILNPRHSVDFKQSRLSEDGLHREGKPINLSHREDLAYMNHIMVSVSPPEEHAMPIGRIADCRLLWDYVYQLLSDSRYENFIRWEDKESKIFRIVDPNGLARLWGNHKNRTNMTYEKMSRALRHYYKLNIIRKEPGQRLLFRFMKTPDEIMSGRTDRLEHLESQELDEQIYQEDECSpecifications
| ETV6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant ETV6. | |
| ETV6 | |
| TEL/TEL/ABL | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC043399 | |
| ETV6 | |
| ETV6 (AAH43399, 1 a.a. ∼ 452 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 2120 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.