missing translation for 'onlineSavingsMsg'
Learn More

exostoses (multiple) 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16147844
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16147844 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16147844 Supplier Abnova Supplier No. H00002131D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human EXT1 protein.

This gene encodes an endoplasmic reticulum-resident type II transmembrane glycosyltransferase involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type I form of multiple exostoses. [provided by RefSeq

Sequence: MQAKKRYFILLSAGSCLALLFYFGGLQFRASRSHSRREEHSGRNGLHHPSPDHFWPRFPDALRPFVPWDQLENEDSSVHISPRQKRDANSSIYKGKKCRMESCFDFTLCKKNGFKVYVYPQQKGEKIAESYQNILAAIEGSRFYTSDPSQACLFVLSLDTLDRDQLSPQYVHNLRSKVQSLHLWNNGRNHLIFNLYSGTWPDYTEDVGFDIGQAMLAKASISTENFRPNFDVSIPLFSKDHPRTGGERGFLKFNTIPPLRKYMLVFKGKRYLTGIGSDTRNALYHVHNGEDVVLLTTCKHGKDWQKHKDSRCDRDNTEYEKYDYREMLHNATFCLVPRGRRLGSFRFLEALQAACVPVMLSNGWELPFSEVINWNQAAVIGDERLLLQIPSTIRSIHQDKILALRQQTQFLWEAYFSSVEKIVLTTLEIIQDRIFKHISRNSLIWNKHPGGLFVLPQYSSYLGDFPYYYANLGLKPPSKFTAVIHAVTPLVSQSQPVLKLLVAAAKSQYCAQIIVLWNCDKPLPAKHRWPATAVPVVVIEGESKVMSSRFLPYDNIITDAVLSLDEDTVLSTTEVDFAFTVWQSFPERIVGYPARSHFWDNSKERWGYTSKWTNDYSMVLTGAAIYHKYYHYLYSHYLPASLKNMVDQLANCEDILMNFLVSAVTKLPPIKVTQKKQYKETMMGQTSRASRWADPDHFAQRQSCMNTFASWFGYMPLIHSQMRLDPVLFKDQVSILRKKYRDIERL

Specifications

Antigen exostoses (multiple) 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human EXT1 protein.
Formulation PBS with no preservative; pH 7.4
Gene EXT1
Gene Accession No. NM_000127
Gene Alias EXT/LGCR/LGS/TRPS2/ttv
Gene Symbols EXT1
Host Species Rabbit
Immunogen EXT1 (NP_000118.2, 1 a.a. ∼ 746 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 2131
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.