Learn More
anti-F13A1, Human, Matched Antibody Pair, Abnova™
Product Code. 16152014
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16152014 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16152014 Supplier Abnova Supplier No. H00002162AP52.1Set
Includes
Capture antibody: mouse monoclonal anti-F13A1, IgG2a Kappa (300μg) , Detection antibody: rabbit polyclonal anti-F13A1 (50μL) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen F13A1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human F13A1.
Gene F13A1
Gene Alias F13A
Gene Symbols F13A1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2162
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-F13A1, IgG2a Kappa (300 ug) 2. Detection antibody: rabbit polyclonal anti-F13A1 (50 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid