missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAAH, Mouse, Clone: 7C9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FAAH.
Brand: Abnova H00002166-M05.100ug
This item is not returnable.
View return policy
Description
This gene encodes a protein that is responsible for the hydrolysis of a number of primary and secondary fatty acid amides, including the neuromodulatory compounds anandamide and oleamide. [provided by RefSeq
Sequence: DLNAPGRATGAVSYTMLYNCLDFPAGVVPVTTVTAEDEAQMEHYRGYFGDIWDKMLQKGMKKSVGLPVAVQCVALPWQEELCLRFMREVERLMTPEKQSSSpecifications
| FAAH | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001441 | |
| FAAH | |
| FAAH (NP_001432, 480 a.a. ∼ 579 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 7C9 | |
| Mouse monoclonal antibody raised against a partial recombinant FAAH. | |
| FAAH | |
| FAAH-1/MGC102823/MGC138146 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2166 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction