Learn More
anti-FABP1 Matched Antibody Pair, Ab-2, Abnova
Brand: Abnova H00002168-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-FABP1 (100μg); Mouse polyclonal anti-FABP1 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
ELISA
Specifications
| FABP1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FABP1. | |
| FABPL/L-FABP | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FABP1 (100 μg) 2. Detection antibody: mouse polyclonal anti-FABP1 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| FABP1 | |
| FABP1 | |
| Affinity Purified | |
| RUO | |
| 2168 | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.