missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas apoptotic inhibitory molecule 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human FAIM3 protein.
Brand: Abnova H00009214-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MDFWLWPLYFLPVSGALRILPEVKVEGELGGSVTIKCPLPEMHVRIYLCREMAGSGTCGTVVSTTNFIKAEYKGRVTLKQYPRKNLFLVEVTQLTESDSGVYACGAGMNTDRGKTQKVTLNVHSEYEPSWEEQPMPETPKWFHLPYLFQMPAYASSSKFVTRVTTPAQRGKVPPVHHSSPTTQITHRPRVSRASSVAGDKPRTFLPSTTASKISALEGLLKPQTPSYNHHTRLHRQRALDYGSQSGREGQGFHILIPTILGLFLLALLGLVVKRAVERRKALSRRARRLAVRMRALESSQRPRGSPRPRSQNNIYSACPRRARGADAAGTGEAPVPGPGAPLPPAPLQVSESPWLHAPSLKTSCEYVSLYHQPAAMMEDSDSDDYINVPASpecifications
| Fas apoptotic inhibitory molecule 3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human FAIM3 protein. | |
| FAIM3 | |
| TOSO | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005449 | |
| FAIM3 | |
| FAIM3 (NP_005440.1, 1 a.a. ∼ 390 a.a) full-length human protein. | |
| 100 μg | |
| Apoptosis | |
| 9214 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- FAIM3 purified MaxPab rabbit polyclonal antibody (D01P)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction