missing translation for 'onlineSavingsMsg'
Learn More

fibulin 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16187697
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16187697 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16187697 Supplier Abnova Supplier No. H00002192D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human FBLN1 protein.

Fibulin 1 is a secreted glycoprotein that becomes incorporated into a fibrillar extracellular matrix. Calcium-binding is apparently required to mediate its binding to laminin and nidogen. It mediates platelet adhesion via binding fibrinogen. Four splice variants which differ in the 3' end have been identified. Each variant encodes a different isoform, but no functional distinctions have been identified among the four variants. [provided by RefSeq]

Sequence: MERAAPSRRVPLQLLLLGGLALLAAGVDADVLLEACCADGHRMATHQKDCSLPYATESKECRMVQEQCCHSQLEELHCATGISLANEQDRCATPHGDNASLEATFVKRCCHCCLLGRAAQAQGQSCEYSLMVGYQCGQVFRACCVKSQETGDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRDTGDEVVCSCFVGYQLLSDGVSCEDVNECITGSHSCRLGESCINTVGSFRCQRDSSCGTGYELTEDNSCKDIDECESGIHNCLPDFICQNTLGSFRCRPKLQCKSGFIQDALGNCIDINECLSISAPCPIGHTCINTEGSYTCQKNVPNCGRGYHLNEEGTRCVDVDECAPPAEPCGKGHRCVNSPGSFRCECKTGYYFDGISRMCVDVNECQRYPGRLCGHKCENTLGSYLCSCSVGFRLSVDGRSCEDINECSSSPCSQECANVYGSYQCYCRRGYQLSDVDGVTCEDIDECALPTGGHICSYRCINIPGSFQCSCPSSGYRLAPNGSNCQDIDECVTGIHNCSINETCFNIQGGFRCLAFECPENYRRSAATRCERLPCHENRECSKLPLRITYYHLSFPTNIQAPAVVFRMGPSSAVPGDSMQLAITGGNEEGFFTTRKVSPHSGVVALTKPVPEPRDLLLTVKMDLSRHGTVSSFVAKLFIFVSAEL

Specifications

Antigen fibulin 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human FBLN1 protein.
Formulation PBS with no preservative; pH 7.4
Gene FBLN1
Gene Accession No. BC022497.1
Gene Alias FBLN
Gene Symbols FBLN1
Host Species Rabbit
Immunogen FBLN1 (AAH22497.1, 1 a.a. ∼ 683 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2192
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.