missing translation for 'onlineSavingsMsg'
Learn More

F-box and leucine-rich repeat protein 8, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16108086
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16108086 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16108086 Supplier Abnova Supplier No. H00055336D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human FBXL8 protein.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. It shares 78% sequence identity with the mouse protein. [provided by RefSeq]

Sequence: MAEPGEGLPEEVLALIFRHLSLRDRAAAARVCRAWAAAATCSAVWHDTKISCECELEGMLPPYLSACLDHIHNLRLEFEPSRKPSRRAAIELLMVLAGRAPGLRGLRLECRGEKPLFDAGRDVLEAVHAVCGAASQLRHLDLRRLSFTLDDALVLQAARSCPELHSLFLDNSTLVGSVGPGSVLELLEACPRLRALGLHLASLSHAILEALAAPDRAPFALLALRCACPEDARASPLPNEAWVALRRRHPGLAVELELEPALPAESVTRVLQPAVPVAALRLNLSGDTVGPVRFAAHHYAATLCALEVRAAASAELNAALEELAARCAALREVHCFCVVSHSVLDAFRAHCPRLRTYTLKLTREPHPWRPTLVA

Specifications

Antigen F-box and leucine-rich repeat protein 8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human FBXL8 protein.
Formulation PBS with no preservative; pH 7.4
Gene FBXL8
Gene Accession No. NM_018378.2
Gene Alias FBL8/FLJ11278/MGC19959
Gene Symbols FBXL8
Host Species Rabbit
Immunogen FBXL8 (NP_060848.2, 1 a.a. ∼ 374 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Primary or Secondary Primary
Gene ID (Entrez) 55336
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.