missing translation for 'onlineSavingsMsg'
Learn More

F-box protein 33, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16122597
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16122597 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16122597 Supplier Abnova Supplier No. H00254170A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant FBXO33.

Members of the F-box protein family, such as FBXO33, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: RNNRNLQKFSLFGDISVLQQQGSLSNTYLSKVDPDGKKIKQIQQLFEEILSNSRQLKWLSCGFMLEIVTPTSLSSLSNAVANTMEHLSLLDNNIPGNSTL

Specifications

Antigen F-box protein 33
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXO33.
Formulation 50% glycerol
Gene FBXO33
Gene Accession No. NM_203301
Gene Alias Fbx33/c14_5247
Gene Symbols FBXO33
Host Species Mouse
Immunogen FBXO33 (NP_976046, 197 a.a. ∼ 296 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 254170
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.