Learn More
F-box protein 9, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FBXO9 protein.
Brand: Abnova H00026268-B01.50uL
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates at least 3 transcript variants diverging at the 5' terminus. [provided by RefSeq]
Sequence: MAEAEEDCHSDTVRADDDEENESPAETDLQAQLQMFRAQWMFELAPGVSSSNLENRPCRAARGSLQKTSADTKGKQEQAKEEKARELFLKAVEEEQNGALYEAIKFYRRAMQLVPDIEFKITYTRSPDGDGVGNSYIEDNDDDSKMADLLSYFQQQLTFQESVLKLCQPELESSQIHISVLPMEVLMYIFRWVVSSDLDLRSLEQLSLVCRGFYICARDPEIWRLACLKVWGRSCIKLVPYTSWREMFLERPRVRFDGVYISKTTYIRQGEQSLDGFYRAWHQVEYYRYIRFFPDGHVMMLTTPEEPQSIVPRLRTRNTRTDAILLGHYRLSQDTDNQTKVFAVITKKKEEKPLDYKYRYFRRVPVQEADQSFHVGLQLCSSGHQRFNKLIWIHHSCHITYKSTGETAVSAFEIDKMYTPLFFARVRSYTAFSERPLSpecifications
| F-box protein 9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FBXO9 protein. | |
| FBXO9 | |
| DKFZp434C0118/FBX9/KIAA0936/NY-REN-57/VCIA1/dJ341E18.2 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_033480.1 | |
| FBXO9 | |
| FBXO9 (NP_258441.1, 1 a.a. ∼ 437 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 26268 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.