Learn More
flap structure-specific endonuclease 1, Mouse, Clone: 1E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FEN1.
Brand: Abnova H00002237-M01A.200uL
Description
The protein encoded by this gene removes 5' overhanging flaps in DNA repair and processes the 5' ends of Okazaki fragments in lagging strand DNA synthesis. Direct physical interaction between this protein and AP endonuclease 1 during long-patch base excision repair provides coordinated loading of the proteins onto the substrate, thus passing the substrate from one enzyme to another. The protein is a member of the XPG/RAD2 endonuclease family and is one of ten proteins essential for cell-free DNA replication. DNA secondary structure can inhibit flap processing at certain trinucleotide repeats in a length-dependent manner by concealing the 5' end of the flap that is necessary for both binding and cleavage by the protein encoded by this gene. Therefore, secondary structure can deter the protective function of this protein, leading to site-specific trinucleotide expansions. [provided by RefSeq]
Sequence: MGIQGLAKLIADVAPSAIRENDIKSYFGRKVAIDASMSIYQFLIAVRQGGDVLQNEEGETTSHLMGMFYRTIRMMENGIKPVYVFDGKPPQLKSGELAKRSERRAEAEKQSpecifications
| flap structure-specific endonuclease 1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_004111 | |
| FEN1 | |
| FEN1 (NP_004102, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA, Western Blot | |
| 1E2 | |
| Mouse monoclonal antibody raised against a partial recombinant FEN1. | |
| FEN1 | |
| FEN-1/MF1/RAD2 | |
| Mouse | |
| 200 μL | |
| Yes | |
| 2237 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.