missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FEZ1 Matched Antibody Pair, Abnova™
Brand: Abnova H00009638-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FEZ1 (100μg); Detection antibody: mouse purified polyclonal anti-FEZ1 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FEZ1
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair
Specifications
| FEZ1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FEZ1. | |
| FEZ1 | |
| Affinity Purified | |
| RUO | |
| 9638 | |
| Human |
| ELISA | |
| Unconjugated | |
| FEZ1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FEZ1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-FEZ1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction