Learn More
FGF1, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00002246-PW2.1Set
Product Code. 16142044
  • € 843.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-FGF1 (300μg), Antibody pair for WB: rabbit polyclonal anti-FGF1 (50μL)
Specifications
FGF1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
AFGF/ECGF/ECGF-beta/ECGFA/ECGFB/FGF-alpha/FGFA/GLIO703/HBGF1
Mouse, Rabbit
RUO
2246
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
FGF1
FGF1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-FGF1 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-FGF1 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.