Learn More
fibroblast growth factor 18, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FGF18 protein.
Brand: Abnova H00008817-B01P.50ug
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures. [provided by RefSeq
Sequence: MYSAPSACTCLCLHFLLLCFQVQVLVAEENVDFRIHVENQTRARDDVSRKQLRLYQLYSRTSGKHIQVLGRRISARGEDGDKYAQLLVETDTFGSQVRIKGKETEFYLCMNRKGKLVGKPDGTSKECVFIEKVLENNYTALMSAKYSGWYVGFTKKGRPRKGPKTRENQQDVHFMKRYPKGQPELQKPFKYTTVTKRSRRIRPTHPASpecifications
| fibroblast growth factor 18 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FGF18 protein. | |
| FGF18 | |
| FGF-18/ZFGF5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003862 | |
| FGF18 | |
| FGF18 (NP_003853.1, 1 a.a. ∼ 207 a.a) full-length human protein. | |
| 50 μg | |
| Cell Cycle | |
| 8817 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.