missing translation for 'onlineSavingsMsg'
Learn More

fibrinogen-like 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human FGL2 protein.

Brand:  Abnova H00010875-D01P.100ug

Product Code. 16103306

  • € 365.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a secreted protein that is similar to the beta- and gamma-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. This protein may play a role in physiologic functions at mucosal sites. [provided by RefSeq

Sequence: MKLANWYWLSSAVLATYGFLVVANNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPGEVGDNRVRELESEVNKLSSELKNAKEEINVLHGRLEKLNLVNMNNIENYVDSKVANLTFVVNSLDGKCSKCPSQEQIQSRPVQHLIYKDCSDYYAIGKRSSETYRVTPDPKNSSFEVYCDMETMGGGWTVLQARLDGSTNFTRTWQDYKAGFGNLRREFWLGNDKIHLLTKSKEMILRIDLEDFNGVELYALYDQFYVANEFLKYRLHVGNYNGTAGDALRFNKHYNHDLKFFTTPDKDNDRYPSGNCGLYYSSGWWFDACLSANLNGKYYHQKYRGVRNGIFWGTWPGVSEAHPGGYKSSFKEAKMMIRPKHFKP
Specifications

Specifications

fibrinogen-like 2
Polyclonal
Rabbit polyclonal antibody raised against a full-length human FGL2 protein.
FGL2
T49/pT49
Rabbit
Affinity Purified
RUO
10875
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_006682.2
FGL2
FGL2 (NP_006673.1, 1 a.a. ∼ 439 a.a) full-length human protein.
100 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.