Learn More
FGR, Mouse, Clone: 3F2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FGR.
Brand: Abnova H00002268-M08.100ug
Description
This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this gene. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MGCVFCKKLEPVATAKEDAGLEGDFRSYGAADHYGPDPTKARPASSFAHIPNYSNFSSQAINPGFLDSGTIRGVSGIGVTLFIALYDYEASpecifications
| FGR | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC064382 | |
| FGR | |
| FGR (AAH64382, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 3F2 | |
| Mouse monoclonal antibody raised against a partial recombinant FGR. | |
| FGR | |
| FLJ43153/MGC75096/SRC2/c-fgr/c-src2/p55c-fgr/p58c-fgr | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2268 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.