Learn More
fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant TTC11.
Brand: Abnova H00051024-A01.50uL
Description
The balance between fission and fusion regulates the morphology of mitochondria. TTC11 is a component of a mitochondrial complex that promotes mitochondrial fission (James et al., 2003 [PubMed 12783892]).[supplied by OMIM
Sequence: MEAVLNELVSVEDLLKFEKKFQSEKAAGSVSKSTQFEYAWCLVRSKYNDDIRKGIVLLEELLPKGSKEEQRDYVFYLAVGNYRLKEYEKALKYVRGLLQTEPQNNQAKELERLIDKAMKKDGLVGMAIVGGMALGVAGLAGLIGLAVSKSKSSpecifications
| fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant TTC11. | |
| FIS1 | |
| CGI-135/TTC11 | |
| Mouse | |
| 50 μL | |
| Apoptosis | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC003540 | |
| FIS1 | |
| TTC11 (AAH03540.1, 1 a.a. ∼ 152 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 51024 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.