missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAST kinase domains 5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FLJ13149.
Brand: Abnova H00060493-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: NVADKSGAMEMAGLCPAACMQTPRMKLAVQFTNRNQYCYGSRDLLGLHNMKRRQLARLGYRVVELSYWEWLPLLKRTRLEKLAFLHEKVFTSALSpecifications
| FAST kinase domains 5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FLJ13149. | |
| FASTKD5 | |
| FLJ13149/FLJ58294/dJ1187M17.5 | |
| Mouse | |
| 50 μL | |
| Apoptosis | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021826 | |
| FASTKD5 | |
| FLJ13149 (NP_068598, 671 a.a. ∼ 764 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 60493 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction