missing translation for 'onlineSavingsMsg'
Learn More

coiled-coil domain containing 88B, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16169833
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16169833 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16169833 Supplier Abnova Supplier No. H00283234B01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human FLJ37970 protein.

Sequence: MRPWQAGSGGNSAQGSRWGEALSHSALGTPLGNDSDSAIQAPWGRPSPTAKDLVWDGRTPLRPCRNTKQMPTERALRYRNRRNVPSPHPSASDTVGTAGLGVQPSRHWSVSGGPRQPKSSGSQGPQGESLDKEAWALRSSTVSAGARRWSWDECVDRGDGWPPRAAPGWSSGSSRWLPLRQRSLGDPPAEGGWQELAREPPALSRWEAESQCWGTVAWADLEP

Specifications

Antigen coiled-coil domain containing 88B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FLJ37970 protein.
Formulation No additive
Gene CCDC88B
Gene Accession No. BC030194
Gene Alias BRLZ/CCDC88/DKFZp434G0920/FLJ00354/FLJ37970/HkRP3
Gene Symbols CCDC88B
Host Species Mouse
Immunogen FLJ37970 (AAH30194, 1 a.a. ∼ 223 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 283234
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.