Learn More
anti-FMC63, scFv Antibody, Clone: Y45, Precision Avi, ACROBiosystems
HEK293 Monoclonal Antibody
Brand:  ACROBiosystems FM3-BY54-25tests
 View more versions of this product
Product Code. 17256210
  • € 379.00 / 25 tests
Description
Description
Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.
Specifications
Specifications
FMC63
Monoclonal
Biotin
Human (HEK293)
RUO
Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.
Anti-idiotypic Antibodies
IgG1 κ
Flow Cytometry
Y45
Lyophilized from 0.22μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.
25 Tests
Primary
-20°C
Powder