Learn More
FMC7 Mouse anti-Human, V450, Clone: FMC7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 644493
Description
FMC7 recognizes a 105kDa membrane glycoprotein expressed on a subset of B lymphocytes.More than 50% of the peripheral B lymphocytes of normal adults carry the FMC7 antigen at variable density. FMC7-positive B cells are more mature, and they include the subpopulation that responds in vitro to mitogens or antigens. The FMC7 antigen is found on B-cell malignancies of most differentiated stages, such as mantle cell lymphoma, but not in most cases of chronic lymphocytic leukemia (CLL).
Host Species: Mouse
Clone: FMC7
Isotype: IgM, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
| FMC7 | |
| Monoclonal | |
| 100μg/mL | |
| HEPES buffer with gelatin and 0.1% sodium azide. | |
| Human B-lymphoblastoid cell line HRIK | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| FMC7 | |
| Violet Fluor 450 (V450) | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.