Learn More
FOXO1A, Mouse, Clone: 4D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXO1A.
Brand: Abnova H00002308-M05.100ug
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. [provided by RefSeq
Sequence: TQVKTPVQVPLPHPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESIIRNDLMDGDTLDFNFDNVLPNQSFPHSVKTTTHSWVSGSpecifications
| FOXO1A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002015 | |
| FOXO1 | |
| FOXO1A (NP_002006, 556 a.a. ∼ 655 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4D9 | |
| Mouse monoclonal antibody raised against a partial recombinant FOXO1A. | |
| FOXO1 | |
| FKH1/FKHR/FOXO1A | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2308 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.