missing translation for 'onlineSavingsMsg'
Learn More

FRS2, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16177398
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16177398 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16177398 Supplier Abnova Supplier No. H00010818D01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human FRS2 protein.

Sequence: MGSCCSCPDKDTVPDNHRNKFKVINVDDDGNELGSGIMELTDTELILYTRKRDSVKWHYLCLRRYGYDSNLFSFESGRRCQTGQGIFAFKCARAEELFNMLQEIMQNNSINVVEEPVVERNNHQTELEVPRTPRTPTTPGFAAQNLPNGYPRYPSFGDASSHPSSRHPSVGSARLPSVGEESTHPLLVAEEQVHTYVNTTGVQEERKNRTSVHVPLEARVSNAESSTPKEEPSSIEDRDPQILLEPEGVKFVLGPTPVQKQLMEKEKLEQLGRDQVSGSGANNTEWDTGYDSDERRDAPSVNKLVYENINGLSIPSASGVRRGRLTSTSTSDTQNINNSAQRRTALLNYENLPSLPPVWEARKLSRDEDDNLGPKTPSLNGYHNNLDPMHNYVNTENVTVPASAHKIEYSRRRDCTPTVFNFDIRRPSLEHRQLNYIQVDLEGGSDSDNPQTPKTPTTPLPQTPTRRTELYAVIDIERTAAMSNLQKALPRDDGTSRKTRHNSTDLPMLAWK

Specifications

Antigen FRS2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human FRS2 protein.
Formulation No additive
Gene FRS2
Gene Accession No. BC021562
Gene Alias FRS2A/FRS2alpha/SNT/SNT-1/SNT1
Gene Symbols FRS2
Host Species Rabbit
Immunogen FRS2 (AAH21562.1, 1 a.a. ∼ 512 a.a) full-length human protein.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10818
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.