Learn More
FYN, Mouse, Clone: 2G2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FYN.
Brand: Abnova H00002534-M04.100ug
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist. [provided by RefSeq
Sequence: MGCVQCKDKEATKLTEERDGSLNQSSGYRYGTDPTPQHYPSFGVTSIPNYNNFHAAGGQGLTVFGGVNSSSHTGTLRTRGGTGVTLFVALSpecifications
| FYN | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032496 | |
| FYN | |
| FYN (AAH32496, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2G2 | |
| Mouse monoclonal antibody raised against a partial recombinant FYN. | |
| FYN | |
| MGC45350/SLK/SYN | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2534 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.