Learn More
G-CSF Rat anti-Human, Biotin, Clone: BVD11-37G10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554670
Description
The BVD11-37G10 antibody reacts with human granulocyte-colony stimulating factor (G-CSF). The immunogen used to generate the BVD11-37G10 hybridoma was recombinant human G-CSF. The BVD11-37G10 antibody is a neutralizing antibody.
Host Species: Rat
Clone: BVD11-37G10
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant Human G-CSF
ELISA
Specifications
| G-CSF | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human G-CSF | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2a |
| ELISA | |
| BVD11-37G10 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.