missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523189
Additional Details : Weight : 0.01000kg
Description
GRK5 Polyclonal antibody specifically detects GRK5 in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GRK5 | |
Polyclonal | |
Unconjugated | |
GRK5 | |
1200003L19Rik; Alpha-CP4; AlphaCP-4; FP2025; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; Gprk5; GRK; Grk5; GRK5 protein kinase; GRK-pan; Pan-GRK; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11476, 14773, 236300, 281801, 2869, 59075, 59092 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
P34947, P43249, Q62833, Q8VEB1 | |
GRK5 | |
A portion of amino acid 350-400 of human GRK5 | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |