Learn More
GAL, Mouse, Clone: 3C1-G5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GAL.
Brand: Abnova H00051083-M01A.200uL
Description
Galanin is small neuropeptide that functions as a cellular messenger within the central and peripheral nervous systems, modulating diverse physiologic functions (Mechenthaler, 2008 [PubMed 18500643]).[supplied by OMIM
Sequence: MARGSALLLASLLLAAALSASAGLWSPAKEKRGWTLNSAGYLLGPHAVGNHRSFSDKNGLTSKRELRPEDDMKPGSFDRSIPENNIMRTIIEFLSFLHLKEAGALDRLLDLPAAASSEDIERSSpecifications
| GAL | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC030241 | |
| GAL | |
| GAL (AAH30241, 1 a.a. ∼ 123 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51083 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3C1-G5 | |
| Mouse monoclonal antibody raised against a full-length recombinant GAL. | |
| GAL | |
| GALN/GLNN/GMAP/MGC40167 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.