Learn More
anti-GALNS, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00002588-PW1.1Set
Product Code. 16015378
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-GALNS (300μL), Antibody pair for WB: rabbit polyclonal anti-GALNS (50μL)
Specifications
GALNS
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FLJ17434/FLJ42844/FLJ98217/GALNAC6S/GAS/MPS4A
Rabbit
RUO
2588
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
GALNS
GALNS
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-GALNS (300 ul) 2. Antibody pair for WB: rabbit polyclonal anti-GALNS (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.