Learn More
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GALNT3 protein.
Brand: Abnova H00002591-B01P.50ug
Description
This gene encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein encoded by this gene is highly homologous to other family members, however the enzymes have different substrate specificities. [provided by RefSeq
Sequence: MERNMKNKNKMLDLMLEAVNNIKDAMPKMQIGAPVRQNIDAGERPCLQGYYTAAELKPVLDRPPQDSNAPGASGKAFKTTNLSVEEQKEKERGEAKHCFNAFASDRISLHRDLGPDTRPPEYVEEYLLFILYHQALQGREGSpecifications
| UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human GALNT3 protein. | |
| GALNT3 | |
| DKFZp686C10199/GalNAc-T3/HFTC/HHS/MGC61909 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 2591 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC056246 | |
| GALNT3 | |
| GALNT3 (AAH56246.1, 1 a.a. ∼ 141 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.