missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA binding protein 3, Mouse, Clone: 1C1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GATA3.
Brand: Abnova H00002625-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LGSHHTASPWNLSPFSKTSIHHGSPGPLSVYPPASSSSLSGGHASPHLFTFPPTPPKDVSPDPSLSTPGSAGSARQDEKECLKYQVPLPDSMKLESSHSpecifications
| GATA binding protein 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001002295 | |
| GATA3 | |
| GATA3 (NP_001002295.1, 103 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1C1 | |
| Mouse monoclonal antibody raised against a partial recombinant GATA3. | |
| GATA3 | |
| HDR/MGC2346/MGC5199/MGC5445 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2625 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction