missing translation for 'onlineSavingsMsg'
Learn More

GATA3 Mouse, Brilliant Violet 421, Clone: L50-823, BD

Product Code. 15832719
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15832719 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15832719 Supplier BD Biosciences Supplier No. 563349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GATA3 (GATA binding protein 3) is a member of the GATA family of transcription factors. This ∽50-kDa nuclear protein regulates the development and subsequent maintenance of multiple tissues. GATA3 is involved in the development of T lymphocytes (regulates T cell receptor subunit gene expression) and the differentiation of mature T cells to become Th2 cells. The expressed levels of normal or mutant GATA3 are also associated with the behaviors of various cancer cells including estrogen receptor-positive breast carcinoma cells.

The L50-823 monoclonal antibody recognizes human and mouse GATA3.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA3
Applications Flow Cytometry
Classification Monoclonal
Clone L50-823
Conjugate Brilliant Violet 421
Description Gata-3; GATA binding protein 3; GATA-binding factor 3; HDR
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Conserved peptide between trans-activation and DNA-binding domains of human, mouse and rat GATA3
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.