missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA zinc finger domain containing 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GATAD1.
Brand: Abnova H00057798-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: KQSKQEIHRRSARLRNTKYKSAPAAEKKVSTKGKGRRHIFKLKNPIKAPESVSTIITAESIFYKGVYYQIGDVVSVIDEQDGKPYYAQIRGFIQDQYCEKSAALTWLIPSpecifications
| GATA zinc finger domain containing 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GATAD1. | |
| GATAD1 | |
| FLJ22489/FLJ40695/ODAG/RG083M05.2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 57798 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021167 | |
| GATAD1 | |
| GATAD1 (NP_066990, 82 a.a. ∼ 190 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction