missing translation for 'onlineSavingsMsg'
Learn More

GUCY1B2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA536523

Additional Details : Weight : 0.00910kg

Product Code. 15533911

  • € 463.00 / 100µL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.

GUCY1B2 Polyclonal antibody specifically detects GUCY1B2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

GUCY1B2
Polyclonal
Unconjugated
GUCY1B2
B930025B03; GCS beta2; GC-SB2; GCS-beta2; GCS-beta-2; guanylate cyclase 1 soluble subunit beta 2 (pseudogene); guanylate cyclase 1, soluble, beta 2; Guanylate cyclase soluble beta 2 (GTP pyrophosphate - lyase); guanylate cyclase soluble subunit beta-2; Guanylate cyclase, soluble, beta 2 (GTP pyrophosphate - lyase); Gucy1b2; Gucy1b2a; Gucy1b2b; SGC; soluble guanylate cyclase beta 2a; soluble guanylate cyclase beta 2b; soluble guanylyl cyclase beta2 subunit
Rabbit
Antigen affinity chromatography
RUO
239134, 25206, 2974
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O75343, P22717
GUCY1B2
Synthetic peptide corresponding to amino acids 71-120 of human GCS-beta-2
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers