Learn More
GDF2 (N-terminus), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GDF2.
Brand: Abnova PAB3885.400uL
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons. [provided by RefSeq
Specifications
| GDF2 Ab-1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GDF2. | |
| In PBS (0.09% sodium azide) | |
| BMP-9/BMP9 | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| GDF2 | |
| GDF2 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GDF2. | |
| RUO | |
| 2658 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.