Learn More
gastric intrinsic factor (vitamin B synthesis), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GIF protein.
Brand: Abnova H00002694-D01P.100ug
Description
This gene is a member of the cobalamin transport protein family. It encodes a glycoprotein secreted by parietal cells of the gastric mucosa and is required for adequate absorption of vitamin B12. Vitamin B12 is necessary for erythrocyte maturation and mutations in this gene may lead to congenital pernicious anemia. [provided by RefSeq
Sequence: MAWFALYLLSLLWATAGTSTQTQSSCSVPSAQEPLVNGIQVLMENSVTSSAYPNPSILIAMNLAGAYNLKAQKLLTYQLMSSDNNDLTIGQLGLTIMALTSSCRDPGDKVSILQRQMENWAPSSPNAEASAFYGPSLAILALCQKNSEATLPIAVRFAKTLLANSSPFNVDTGAMATLALTCMYNKIPVGSEEGYRSLFGQVLKDIVEKISMKIKDNGIIGDIYSTGLAMQALSVTPEPSKKEWNCKKTTDMILNEIKQGKFHNPMSIAQILPSLKGKTYLDVPQVTCSPDHEVQPTLPSNPGPGPTSASNITVIYTINNQLRGVELLFNETINVSVKSGSVLLVVLEEAQRKNPMFKFETTMTSWGLVVSSINNIAENVNHKTYWQFLSGVTPLNEGVADYIPFNHEHITANFTQYSpecifications
| gastric intrinsic factor (vitamin B synthesis) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human GIF protein. | |
| GIF | |
| IF/IFMH/INF/TCN3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2694 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005142.2 | |
| GIF | |
| GIF (NP_005133.2, 1 a.a. ∼ 417 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.