missing translation for 'onlineSavingsMsg'
Learn More

GJB1, Mouse, Clone: 2B9, Abnova™

Product Code. 16158674
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16158674 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16158674 Supplier Abnova Supplier No. H00002705M03.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant GJB1.

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: RLWSLQLILVSTPALLVAMHVAHQQHIEKKMLRLEGHGDPLHLEEVKRHKVHISGTLWWTYVISVVFRLLFEAVFMYVFYLLYPGYAMVRLVKCDVYPCP

Specifications

Antigen GJB1
Applications ELISA
Classification Monoclonal
Clone 2B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GJB1.
Formulation PBS with no preservative; pH 7.4
Gene GJB1
Gene Accession No. BC022426
Gene Alias CMTX/CMTX1/CX32
Gene Symbols GJB1
Host Species Mouse
Immunogen GJB1 (AAH22426, 75 a.a. ∼ 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2705
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.