missing translation for 'onlineSavingsMsg'
Learn More
Learn More
guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GNAL.
Brand: Abnova H00002774-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: DMLAEKVLAGKSKIEDYFPEYANYTVPEDATPDAGEDPKVTRAKFFIRDLFLRISTATGDGKHYCYPHFTCAVDTENIRRVFNDCRDIIQRMHLKQYELLSpecifications
| guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GNAL. | |
| GNAL | |
| GNAL | |
| GNAL (NP_892023, 359 a.a. ∼ 458 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 2774 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_182978 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction