Learn More
GREM1, Mouse, Clone: 2C8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GREM1.
Brand: Abnova H00026585-M02A.200uL
Description
This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to relay the sonic hedgehog (SHH) signal from the polarizing region to the apical ectodermal ridge during limb bud outgrowth. [provided by RefSeq
Sequence: ESSQEALHVTERKYLKRDWCKTQPLKQTIHEEGCNSRTIINRFCYGQCNSFYIPRHIRKEEGSFQSCSFCKPKKFTTMMVTLNCPELQPPTKKKRVTRVKQCRCISIDLDSpecifications
| GREM1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_013372 | |
| GREM1 | |
| GREM1 (NP_037504, 75 a.a. ∼ 184 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 26585 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 2C8 | |
| Mouse monoclonal antibody raised against a partial recombinant GREM1. | |
| GREM1 | |
| CKTSF1B1/DAND2/DRM/GREMLIN/IHG-2/MGC126660/PIG2 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.