missing translation for 'onlineSavingsMsg'
Learn More

anti-HA, Clone: 1A9, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova MAB4953-M10.100ug

Product Code. 16173047

  • € 410.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HA
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
Mouse
100 μg
Primary
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
ELISA, Western Blot
1A9
Mouse monoclonal antibody raised against partial recombinant HA (Influenza A virus H7N1).
CY024754.1
Recombinant Fc fusion protein corresponding to amino acids 21-523 virus of HA (Influenza A virus, A/turkey/Italy/977/1999 (H7N1))
RUO
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only