missing translation for 'onlineSavingsMsg'
Learn More

anti-HA, Clone: 4B9, Abnova™

Product Code. 16183047
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16183047 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16183047 Supplier Abnova Supplier No. MAB6542M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen HA
Applications ELISA
Classification Monoclonal
Clone 4B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant HA (Influenza A virus H5N1).
Formulation In 1x PBS, pH 7.4
Gene Accession No. AY651336.1
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to amino acids 17-531 of virus HA (Influenza A virus, A/Viet Nam/3062/2004 (H5N1))
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.