missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAGH, Mouse, Clone: 2F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HAGH.
Brand: Abnova H00003029-M02A.200uL
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. [provided by RefSeq
Sequence: GRLPPDTRVYCGHEYTINNLKFARHVEPGNAAIREKLAWAKEKYSIGEPTVPSTLAEEFTYNPFMRVREKTVQQHAGETDPVTTMRAVRREKDQFKMPRDSpecifications
| HAGH | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005326 | |
| HAGH | |
| HAGH (NP_005317, 161 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3029 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2F9 | |
| Mouse monoclonal antibody raised against a partial recombinant HAGH. | |
| HAGH | |
| GLO2/GLX2/GLXII/HAGH1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido