missing translation for 'onlineSavingsMsg'
Learn More

anti-Hamster IgG2, κ, Brilliant Violet 650, Clone: B81-3, BD

Brand:  BD Biosciences 563860

Product Code. 15873159

  • € 435.00 / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Hamster
Clone: B81-3
Isotype: IgG2, κ
Immunogen: Keyhole Limpet Hemocyanin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
B81-3
Brilliant Violet 650
Keyhole Limpet Hemocyanin
50 μg
Isotype Control (Primary)
IgG2 κ
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.