Learn More
HSP90 beta Monoclonal Antibody (H90-10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110892
Additional Details : Weight : 0.01000kg
Description
MA1-10892 detects 90 kDa Hsp90 beta in all reactive species except chicken where it detects both alpha and beta isoforms. MA1-10892 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, MA1-10892 is specific for the ∽90 kDa Heat Shock Protein 90 protein. MA1-10892 immunogen is purified human Hsp90 beta.
HSP90 beta Monoclonal antibody specifically detects HSP90 beta in Human, Mouse, Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
HSP90 beta | |
Monoclonal | |
1.3 mg/mL | |
PBS with no preservative; pH 7.4 | |
P08238, P11499, P30947 | |
Hsp90ab1 | |
Purified human Hsp90 beta. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
H90-10 | |
Unconjugated | |
Hsp90ab1 | |
90kDa; 90-kDa heat shock protein beta; AL022974; C81438; D6S182; heat shock 84 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; heat shock cognate protein HSP 90-beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90 kDa alpha class B member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; Hsp84; Hsp84-1; Hsp90; hsp90 beta; HSP90AB1; HSP90B; HSP90-beta; HSPC2; HSPCB; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen; Unknown (protein for IMAGE:7943063) | |
Mouse | |
Protein G | |
RUO | |
100358690, 15516, 3326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |