Learn More
HEY1, Mouse, MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HEY1 protein.
Brand: Abnova H00023462-B02.50uL
Description
This gene encodes a nuclear protein belonging to the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcriptional repressors. Expression of this gene is induced by the Notch and c-Jun signal transduction pathways. Two similar and redundant genes in mouse are required for embryonic cardiovascular development, and are also implicated in neurogenesis and somitogenesis. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: MKRAHPEYSSSDSELDETIEVEKESADENGNLSSALGSMSPTTSSQILARKRRRGIIEKRRRDRINNSLSELRRLVPSAFEKQGSAKLEKAEILQMTVDHLKMLHTAGGKGYFDAHALAMDYRSLGFRECLAEVARYLSIIEGLDASDPLRVRLVSHLNNYASQREAASGAHAGLGHIPWGTVFGHHPHIAHPLLLPQNGHGNAGTTASPTEPHHQGRLGSAHPEAPALRAPPSGSLGPVLPVVTSASKLSPPLLSSVASLSAFPFSFGSFHLLSPNALSPSAPTQAANLGKPYRPWGTEIGAFSpecifications
| HEY1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human HEY1 protein. | |
| HEY1 | |
| BHLHb31/CHF2/HERP2/HESR1/HRT-1/MGC1274/OAF1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_012258.3 | |
| HEY1 | |
| HEY1 (NP_036390.3, 1 a.a. ∼ 304 a.a) full-length human protein. | |
| RUO | |
| 23462 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.