Learn More
HEYL, Mouse, Clone: 4A1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HEYL.
Brand: Abnova H00026508-M01.100ug
Description
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. [provided by RefSeq
Sequence: PLRRATGIILPARRNVLPSRGASSTRRARPLERPATPVPVAPSSRAARSSHIAPLLQSSSPTPPGPTGSAAYVAVPTPNSSSPGPAGRPAGAMLYHSWVSEITEIGASpecifications
| HEYL | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014571 | |
| HEYL | |
| HEYL (NP_055386, 221 a.a. ∼ 327 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4A1 | |
| Mouse monoclonal antibody raised against a partial recombinant HEYL. | |
| HEYL | |
| HRT3/MGC12623/bHLHb33 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26508 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.