Learn More
HLA-DR, DP, DQ Mouse anti-Human, Unlabeled, Clone: TÜ39, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555557
Description
Reacts with all major histocompatibility Class II HLA-DR, DP and most DQ antigens expressed on B cells, antigen presenting cells and activated T cells. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| HLA-DR, DP, DQ | |
| Monoclonal | |
| 0.5mg/mL | |
| Tü39 | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| TÜ39 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.